D-(+)-Xylose
Bottle, 500 g
Catalog # APOSOR7967-500G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:58-86-6
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00151475
- Molecular Weight:150.13 g/mol
- Melting Pt:147…158 °C
- Density:1.525 g/cm³ (25 °C)
- Flash Pt:>110 °C
- Storage Temperature:Ambient
- EINECS:200-400-7
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:500 g



