Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dichloropropane 99%

1,3-Dichloropropane 99%

Bottle, 100 g
Catalog # APOSOR0973-100G
Flammable
Supplier:   Apollo Scientific
1,3-Dichloropropane 99%
Catalog # APOSOR0973-100G
Supplier:  Apollo Scientific
CAS Number:  142-28-9
1,3-Dichloropropane 99%

Bottle, 100 g

CAS Number:  142-28-9
Price
89,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Trimethylendichloride
  • CAS Number:142-28-9
  • Formula:ClH₂CCH₂CH₂Cl
  • MDL Number:MFCD00000999
  • Molecular Weight:112.99 g/mol
  • Boiling Pt:120 °C
  • Density:1.19
  • Flash Pt:21 °C
  • UN:1933
  • ADR:3,II
  •  :Density: 1.1717 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications