Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dimethoxyphenol

3,5-Dimethoxyphenol

Bottle, 1 g
Catalog # APOSOR1170-1G
Irritant
Supplier:   Apollo Scientific
3,5-Dimethoxyphenol
Catalog # APOSOR1170-1G
Supplier:  Apollo Scientific
CAS Number:  500-99-2
3,5-Dimethoxyphenol

Bottle, 1 g

CAS Number:  500-99-2
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:500-99-2
  • Formula:C₈H₁₀O₃
  • MDL Number:MFCD00008388
  • Molecular Weight:154.17 g/mol
  • Boiling Pt:172…175/17 mm °C
  • Boiling Pt:172 - 175/17 mm °C
  • Melting Pt:40…45 °C
  • Melting Pt:40 - 45 °C
  • Flash Pt:78 °C
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications