Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Difluorothiobenzamide 95%

2,4-Difluorothiobenzamide 95%

Bottle, 100 mg
Catalog # APOSPC32594-100MG
Irritant
Supplier:   Apollo Scientific
2,4-Difluorothiobenzamide 95%
Catalog # APOSPC32594-100MG
Supplier:  Apollo Scientific
CAS Number:  175276-92-3
2,4-Difluorothiobenzamide 95%

Bottle, 100 mg

CAS Number:  175276-92-3
Price
21,80 €
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:175276-92-3
  • Formula:C₇H₅F₂NS
  • MDL Number:MFCD00085011
  • Molecular Weight:173.19 g/mol
  • Melting Pt:128…132 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg

Specifications