2-Methoxy-4-methylphenol 98%
Bottle
Catalog # APOSOR8723-500G
Supplier: Apollo Scientific
2-Methoxy-4-methylphenol 98%
Catalog # APOSOR8723-500G
Supplier: Apollo Scientific
CAS Number: 93-51-6
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol
- CAS Number:93-51-6
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00002378
- Molecular Weight:138,16379 g/mol
Specifications
Specifications
- Pack type:Bottle



