Order Entry
Export
Orders LinkContactUsLinkComponent
2-Methoxy-4-methylphenol 98%

2-Methoxy-4-methylphenol 98%

Bottle
Catalog # APOSOR8723-100G
Supplier:   Apollo Scientific
2-Methoxy-4-methylphenol 98%
Catalog # APOSOR8723-100G
Supplier:  Apollo Scientific
CAS Number:  93-51-6
2-Methoxy-4-methylphenol 98%

Bottle

CAS Number:  93-51-6
Price
60,80 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol
  • CAS Number:93-51-6
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00002378
  • Molecular Weight:138,16379 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications