Propidium Iodide
Bottle, 50 mg
Catalog # APOSBIP4355-50MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Fluorescent staining compound.
Synonyms: 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diiodide, PI, 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide
- CAS Number:25535-16-4
- Formula:C₂₇H₃₄I₂N₄
- MDL Number:MFCD00011921
- Molecular Weight:668.4 g/mol
- Melting Pt:220…225 °C (dec.)
- Storage Temperature:Refrigerator
- EINECS:247-081-0
Specifications
Specifications
- Pack type:Bottle
- Pk:50 mg



