3'-Acetylacetophenone 98%
Bottle
Catalog # APOSOR59987-100G
Supplier: Apollo Scientific
3'-Acetylacetophenone 98%
Catalog # APOSOR59987-100G
Supplier: Apollo Scientific
CAS Number: 6781-42-6
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Benzene-1,3-bis(acetyl)
- CAS Number:6781-42-6
- Formula:C₁₀H₁₀O₂
- MDL Number:MFCD00008740
- Molecular Weight:162,18519 g/mol
Specifications
Specifications
- Pack type:Bottle



