Order Entry
Export
Orders LinkContactUsLinkComponent
4-Methylumbelliferone

4-Methylumbelliferone

Bottle, 25 g
Catalog # APOSOR7969-25G
Irritant
Supplier:   Apollo Scientific
4-Methylumbelliferone
Catalog # APOSOR7969-25G
Supplier:  Apollo Scientific
CAS Number:  90-33-5
4-Methylumbelliferone

Bottle, 25 g

CAS Number:  90-33-5
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Hymecromone, 7-Hydroxy-4-methylcoumarin, 7-Hydroxy-4-methyl-2H-chromen-2-one, β-Methylumbelliferone, 4-MU
  • CAS Number:90-33-5
  • Formula:C₁₀H₈O₃
  • MDL Number:MFCD00006866
  • Molecular Weight:176.17 g/mol
  • Melting Pt:190…192 °C
  • Storage Temperature:Ambient
  • EINECS:201-986-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications