7-Methylindole
Bottle, 100 g
Catalog # APOSOR6655-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:933-67-5
- Formula:C₉H₉N
- MDL Number:MFCD00005684
- Molecular Weight:131.18 g/mol
- Boiling Pt:266 °C
- Melting Pt:80…84 °C
- :Density: 1.0202 g/cm³ (99.6 °C)
- Storage Temperature:Ambient
- EINECS:213-270-1
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



