Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Nitrobenzonitrile 99%

Bottle, 1 g
Catalog # APOSOR5001-1G
Toxic
Supplier:   Apollo Scientific
2-Nitrobenzonitrile 99%
Catalog # APOSOR5001-1G
Supplier:  Apollo Scientific
CAS Number:  612-24-8
2-Nitrobenzonitrile 99%

Bottle, 1 g

CAS Number:  612-24-8
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  o-Nitrobenzonitrile
  • CAS Number:612-24-8
  • Formula:C₇H₄N₂O₂
  • MDL Number:MFCD00007044
  • Molecular Weight:148.12 g/mol
  • Boiling Pt:165 °C/16mmHg
  • Melting Pt:107…110 °C
  • Storage Temperature:Ambient
  • EINECS:210-301-0
  • UN:3439
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications