Order Entry
Export
Orders LinkContactUsLinkComponent
Capsaicin

Capsaicin

Bottle, 10 g
Catalog # APOSBIC0125-10G
Supplier:   Apollo Scientific
Capsaicin
Catalog # APOSBIC0125-10G
Supplier:  Apollo Scientific
CAS Number:  404-86-4
Capsaicin

Bottle, 10 g

CAS Number:  404-86-4
Price
232,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Active component of cayenne pepper.

Synonyms:  8-Methyl-N-vanillyl-6-none-amid
  • CAS Number:404-86-4
  • Formula:C₁₈H₂₇NO₃
  • MDL Number:MFCD00017259
  • Molecular Weight:305.42 g/mol
  • Boiling Pt:210…220 °C
  • Melting Pt:65…68 °C
  • Flash Pt:113 °C
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications