Order Entry
Export
Orders LinkContactUsLinkComponent
 

Isopropylurea 97%

Bottle
Catalog # APOSOR54393-1G
Supplier:   Apollo Scientific
Isopropylurea 97%
Catalog # APOSOR54393-1G
Supplier:  Apollo Scientific
CAS Number:  691-60-1
Isopropylurea 97%

Bottle

CAS Number:  691-60-1
Price
76,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:691-60-1
  • Formula:C₄H₁₀N₂O
  • MDL Number:MFCD00047875
  • Molecular Weight:102,135 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications