Isopropylurea 97%
Bottle
Catalog # APOSOR54393-250MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
- CAS Number:691-60-1
- Formula:C₄H₁₀N₂O
- MDL Number:MFCD00047875
- Molecular Weight:102,135 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSOR54393-250MG



