α,2,4-Trichlorobenzaldoxime
Bottle, 250 mg
Catalog # APOSOR29855-250MG
Supplier: Apollo Scientific
α,2,4-Trichlorobenzaldoxime
Catalog # APOSOR29855-250MG
Supplier: Apollo Scientific
CAS Number: 29203-60-9
Jump to:
Product Details & Documents
Synonyms: 2,4-Dichloro-N-hydroxybenzenecarboximidoyl chloride
- CAS Number:29203-60-9
- Formula:C₇H₄Cl₃NO
- MDL Number:MFCD00192643
- Molecular Weight:224.472 g/mol
- Melting Pt:93…95 °C
- Storage Temperature:Ambient
- UN:1759
- ADR:8,III
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



