1H-Pyrazolo[4,3-b]pyridine
Bottle, 100 mg
Catalog # APOSOR30695-100MG
Supplier: Apollo Scientific
1H-Pyrazolo[4,3-b]pyridine
Catalog # APOSOR30695-100MG
Supplier: Apollo Scientific
CAS Number: 272-52-6
Jump to:
Product Details & Documents
Synonyms: Pyrazolo[4,3-b]pyridine
- CAS Number:272-52-6
- Formula:C₆H₅N₃
- MDL Number:MFCD08234560
- Molecular Weight:119.13 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:100 mg



