Order Entry
Export
Orders LinkContactUsLinkComponent
Lead (II) Bromide 99,9%

Lead (II) Bromide 99,9%

Bottle
Catalog # APOSIN2208-500G
Irritant
Environment
Health
Supplier:   Apollo Scientific
Lead (II) Bromide 99,9%
Catalog # APOSIN2208-500G
Supplier:  Apollo Scientific
CAS Number:  10031-22-8
Lead (II) Bromide 99,9%

Bottle

CAS Number:  10031-22-8
Price
319,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Inorganic salt or catalyst.

Synonyms:  Lead dibromide, Lead(II) bromide
  • CAS Number:10031-22-8
  • Formula:Pb·2Br
  • MDL Number:MFCD00011156
  • Molecular Weight:367,008 g/mol
  • EINECS:233-084-4
  • UN:2291
Specifications

Specifications

  • Pack type:
    Bottle

Specifications