Lead (II) Bromide 99,9%
Bottle
Catalog # APOSIN2208-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Inorganic salt or catalyst.
Synonyms: Lead dibromide, Lead(II) bromide
- CAS Number:10031-22-8
- Formula:Pb·2Br
- MDL Number:MFCD00011156
- Molecular Weight:367,008 g/mol
- EINECS:233-084-4
- UN:2291
Specifications
Specifications
- Pack type:Bottle



