Orders LinkContactUsLinkComponent
VWR®, Macro Pipette Tips
VWR®, Macro Pipette Tips
Macro pipette tips, 5000 - 10000 µl, +, +, 160,8 mm, Clear, VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors, Rack of 25 tips
Catalog # 732-3917
Supplier:   VWR Collection
VWR®, Macro Pipette Tips
Catalog # 732-3917
Supplier:  VWR Collection
VWR®, Macro Pipette Tips
VWR®, Macro Pipette Tips
Macro pipette tips, 5000 - 10000 µl, +, +, 160,8 mm, Clear, VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors, Rack of 25 tips
Price
42,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Specifications
  • Packed:
    Rack of 25 tips
  • Sterile:
    +
  • Volume:
    5000 - 10000 µl
  • Length:
    160,8 mm
  • Seq:
    12
  • Tip style:
    Macro pipette tips
  • Tip colour:
    Clear
  • For use with:
    VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors
  • Filter:
    +