Orders LinkContactUsLinkComponent
VWR®, Macro Pipette Tips
VWR®, Macro Pipette Tips
Macro pipette tips, 1000 - 5000 µl, -, +, 123,6 mm, Clear, VWR, Gilson, Rainin Classic, and other universal-style pipettors, Bulk, 100 per bag
Catalog # 732-3912
Supplier:   VWR Collection
VWR®, Macro Pipette Tips
Catalog # 732-3912
Supplier:  VWR Collection
VWR®, Macro Pipette Tips
VWR®, Macro Pipette Tips
Macro pipette tips, 1000 - 5000 µl, -, +, 123,6 mm, Clear, VWR, Gilson, Rainin Classic, and other universal-style pipettors, Bulk, 100 per bag
Price
32,70 €
Log in for availability
1 * 100 items
Product Details & Documents
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Specifications
  • Packed:
    Bulk, 100 per bag
  • Sterile:
    -
  • Volume:
    1000 - 5000 µl
  • Length:
    123,6 mm
  • Seq:
    07
  • Tip style:
    Macro pipette tips
  • Tip colour:
    Clear
  • For use with:
    VWR, Gilson, Rainin Classic, and other universal-style pipettors
  • Filter:
    +