0,00 €
Order Entry
/
Products
Overview
Life Science Products
Cell Biology
Sera
Media
Supplements and Antibiotics
Cytokines, Growth Factors and Inhibitors
Transfection
Extracellular Matrices and Coated Plates
Cell Culture Consumables
Antibodies
Immunoassays and Cellular Assays
Cell Biology Equipment and Instruments
Genomics
Nucleic Acid Sample Prep
Cloning and Gene Editing
Sequencing
DNA Electrophoresis
Amplification
Microbiology
Culture Media for Microbiology
Rapid Testing
Microorganism Tests
Staining Solutions
Kits for Microscopy
Hygiene Monitoring
Anaerobic Supplies and Gas Generators
Air Samplers
Colony Counters
Gravimetric Dilutors
Media Preparators
Media Dispensers
Proteomics
Protein Extraction
Protein Purification
Electrophoresis and Blotting
Protein Detection and Quantification
Antibodies
Protein Linking and Labeling
Peptide Synthesis
Equipment & Instruments
Autoclaves and Sterilizers
Balances
Laboratory Balances
Industrial Balances
Moisture Balances
Centrifuges
Mini Centrifuges
Micro Centrifuges
Benchtop Centrifuges
Floor Standing Centrifuges
Environment
Water Samplers
Flocculation
Flow Meters
Turbidity Meters
BOD Meters
Spectrophotometers
Biophotometers
Flame Photometers
Reflectometers
Heating and Cooling
Cryothermostats and Chillers
Heating Circulators and Thermostats
Baths, Water Baths and Shaking Baths
Refrigerators
Freezers
Ultralow Temperature Freezers
Cryogenic Storage
Incubators
Climatic Testing Chambers
Standard Ovens
High Temperature Ovens
Furnaces
Liquid Handling Instruments
Dilutors
Dispensers and Pumps
Mechanical Pipettes
Microbiology
Air Samplers
Laboratory Paddle Blenders
Standard Laboratory Blenders
Media Dispensers
Colony Counters
Microscopy
Microscopes
Cameras
Illumination
Magnifiers
Mixers, Stirrers, Shakers and Grinders
Homogenizers
Mills
Shakers
Stirrers
Vortexers
Grinders
Mortar and Pestles
Physical Measurements
Viscometers
Soxhlet Extractors
Refractometers
Polarimeters
Distillation Systems
Flash Point Testers
Densimeters
Hydrometers
Temperature Probes
Calorimeters
Manometers
Melting Point Apparatus
Pumps
Peristaltic Pumps
Liquid Pumps
Vacuum Pumps
Vacuum Rotary Vane Pumps
Thermometers and Hygrometers
Digital Thermometers
Analog Thermometers
Hygrometers
Chart Recorders
Laboratory Furniture
Work Benches
Gowning Benches
Shelves
Gowning Shelves and Racks
Tables and Workstations
Tables
Weighing Tables
Modular Workstations
Lab Chairs and Stools
Ladders and Step Stools
Fume Hoods and Enclosures
Pharmacy Fume Hoods
Extraction Cabinets
Animal Research Enclosures
Balances Enclosures
Chemical Fume Hoods
Forensic Enclosures
Ductless Fume Hoods
Laminar Flow Cabinets
Carts
Glove Boxes
Laboratory Consumables
Glassware
Culture Tubes
Vials
Bottles
Flasks
Graduated Cylinders
Beakers
Jars
Evaporating Basins and Dishes
Funnels
Cells and Cuvettes
Distillation Glassware
Desiccators
Anatomy, Pathology and Clinical
Slides
Coverslips
Counting Chambers
Syringe Needles
Medical Syringes
Swabs for Clinical Use
Tongue Depressor
Lancets
Blades
Sample Containers
Staining Troughs
Staining Racks
Filtration
Filtration Manifolds
Bottle Top Filter Units
Filtration Capsules
Syringe Filters
Vent Filters
Dialysis Tubing
Membrane Filters
Tangential Cross Flow Devices
Filter Plates
Filter Discs
Filter Papers
Centrifugal Devices
General Plasticware
Carboys
Plastic Bottles
Cell Culture Flasks
Centrifuge Tubes
Tube Racks
Sealing Mats and Film
Plastic Weighing Dishes
Transfer Pipets
Petri Dishes
Deep Well Links
Cryovials
Serological Pipets
Crucibles and Metal Type
Crucibles
Weighing Dishes
Scalpes
Tongs
Forceps and Tweezers
Scissors
Aluminum Closures
Stands
Sieves
Magnetic Stirring Bars
Liquid Handling
Manual Pipette Tips
Robotic Pipette Tips
Reservoirs
Bottle Top Dispensers
Burettes
Mechanical Pipettes
Transfer Pipettes
Automated Liquid Handling Platforms
Steppers and Repetitive Pipettes
Pipette Controllers and Fillers
Electronic Pipettes
Electrochemistry
pH Electrodes
Ion Specific Electrodes
Conducitivity Electrodes
Dissolved Oxygen Electrodes
Redox Electrodes
Chemicals
Chemicals by Brand
Chemical by Methods
HPLC
Titration
pH Measurement
UV/VIS Spectroscopy
Dissolution Testing
Gas Chromatography
Conductivity Measurement
Physical Property Standards
Nuclear Magnet Resonance
AAS/ICP
Nitrogen Determination
Chemicals by Category
Organic Synthesis
Custom Manufacturing Solutions
Chromatography
Sample Preparation
Syringe Filters
Sample Phase Extraction Plates
Automatic Extractor Systems
Sample Preparation Columns
Manual Solid Phase Extraction Unit
Solid Phase Extraction Sorbents
Solid Phase Extraction Columns, Cartridges, Disks
Liquid Chromatography
U/HPLC Instruments
Chromatography Columns
Chromatography Vials
Syringes
Fittings, Tubing and Accessories
Bulk Material
Gas Chromatography (GC)
GC Instruments
Chromatography Software
Chromatography Columns
GC Fittings and Accessories
Chromatography Syringes
Syringe Needles
Gas Generators and Accessories
GC Derivatization Reagents
GC Solvents
TLC/HPTLC
Plates
Equipment and Accessories for TLC
HPLC Solvents
Preparative and Flash Chromatography
Preparative Instruments
Chromatography Software
Preparative/Flash Chromatography Columns
Bulk Sorbent
ACS Solvents
Tubing and Connectors
Flash Instruments
Silicones
Additives
Adhesives and Sealants
Coatings and Dispersions
Conductive Materials
Elastomers
High Consistency Rubbers
Liquid Silicone Rubbers
Low Viscosity Elastomers
Inks
Lubricants
Polymers and Catalysts
Potting and Encapsulating
Primers
Skin Care Ingredients
Healthcare Products
Clinical Chemistry
Analyzers
Drug of Abuse Tests
Serological Tests
Chemistry and Immunoassay
Blood and Specimen Collection and Transport
Blood Collection
Urine Collection
Specimen Collection Containers
Clinical Swabs
Sample Bags
Sample Transport Containers
Cooling Packs
Surgical Drapes
Temperature Indicators
Swab
Blood Collection Tubes
Histology and Cytology
Tissue Processing Consumables
Histology and Anatomic Pathology Equipment
Histology Stains
Microscopy and Staining Reagents
Archiving and Storage
Staining
Hematology
Cell Counters
Hematology Staining
Tube Rotators and Rollers
Medical Syringes
Syringe Needles
Hematology Reagents
Clinical General Equipment
Centrifuges
Microscopes
Cold Storage
Biological Safety Cabinets
Water Purification Systems
Clinical Microbiology
Microbiology Stains
Dehydrated Media
Prepared Media
Microbiology Dyes
CO₂ Incubators
Parasite Testing
Bacterial Test Kits, Viral Test Kits and Controls
Microbiology Consumables
Colony Counters
Clinical Quality Controls
Controlled Environment, Safety & PPE
Gloves and Hand Protection
Laboratory Gloves
Cleanroom Gloves
Chemical-Resistant Gloves
Industrial Gloves
Special Application Gloves
Personal and Product Protection
Eye and Face Protection
Respiratory Protection
Hearing and Head Protection
Disposable Apparel
Reusable Garments and Workwear
Footwear
Skin Protection and Hygiene
Fall Protection
Gas Detection
Radiation Protection
Gas Detection Systems
First Aid and Emergency
First Aid Kits
Bandages and Wound Care
Eye Wash
Emergency Showers
Fire Blankets
Evacuation Equipment
Facility Protection and Cleaning
Spill Control
Cleaning and Disinfection
Waste Management and Recycling
Bench Protection
Mats/Flooring
Electrostatic Discharge Control
Tapes
Lockout/Tagout
Equipment and Accessories
Printers and Labels
Tape and Stationary
Packaging
Covers
Fluid Management
Wafer and Die Handling
Chemical Handling and Storage
Environmental Monitoring
Glove Boxes and Glove Bags
Tools and Industrial Supplies
Abrasives
Adhesives, Sealants and Tapes
Chemical Lubricants
Electrical and Lighting
Power Tools
Tool Sets
Fasteners, Clamps and Straps
Hand Tools
Lab and Permanent Markers
Carts and Trolleys
Safety Cans
Measuring and Leveling Tools
Fluid Management
Fluid Handling Pumps
Masterflex Peristaltic Pumps
Piston Pumps
Gear Pumps
Syringe Pumps
Centrifugal Pumps
Diaphragm Pumps
Drum/Transfer Pumps
HPLC/Gel PC Pumps
Microfluidic Pumps
Tubing Pumps and Hose Pumps
Progressive Cavity Pumps
Rotary Lobe Pumps
Tubing and Fittings
Masterflex Pump Tubing
Transfer Flexible Tubing
Transfer Rigid Tubing
Fittings
Tubing and Sanitary Clamps
Gaskets
Tubing Accessories
Fluid Measurement and Control
Valves
Flow Indicators
Flow Switches
Flow Sensors
Level Sensors
Flow Meters
Single-Use Assemblies & Consumables
Bioprocess Filtration
Fluid Transfer Assemblies
Sampling Assemblies
Storage & Mixing Bag Assemblies
Single-Use Systems
Mixing Systems
Bioprocess Containers
Custom Bioprocess Systems
Bioprocess Ingredients & Excipients
Active Pharmaceutical Ingredients
Excipients
Buffer Management
Hydrated Buffers, Reagents, & Cell Culture Media
J.T.Baker Direct Dispense Packaging System
Flowmor Packaging System
Services
Overview
Laboratory & Production Services
Procurement & Material Management Services
Procurement & Supply Chain Services
Inventory Management Solutions
Chemical Management Solutions
General Lab Services
Technical Lab Services
Scientific Services
Media & Buffer Preparation
Sample Management Services
Production Services
Critical Environment & Sanitization
Production Process Readiness
Planning & Material Management
Downstream Optimization Services
Business Process Consulting
Lean Consulting
Project Management
Procurement & Sourcing
Marketsource Sourcing Solutions
Marketsource Online Product Portfolio
Equipment & Instrument Services
Multi-Vendor Technical Services
Compliance Services
Equipment Service Offerings
Controlled Temperature Units
Balances, Scales and Weighing
Chromatography and Mass Spectroscopy
Fume Hoods and Safety Cabinets
Laboratory Equipment and Instruments
Pipette Calibration & Volumetric Testing
The Journey of a Drug
Downstream Optimization
Digital Solutions
Overview
Workflow Optimization
Software Solutions
Chemical Manager
Equipment Manager
Hardware Solutions
Digital Commerce
Inventory Manager
e-Commerce Tools
B2B Integration Services
Data Informatics
Data, Metrics & Analytic Solutions
Digital Services
Applications
Overview
Bioprocess
Monoclonal Antibodies (mAbs)
Recombinant Proteins
Recombinant Proteins Upstream Processing
Recombinant Proteins Downstream Processing
Recombinant Proteins Formulation and Final Fill
Gene Therapy
Your unique gene therapy at scale
Gene Therapy Upstream Processing
Gene Therapy Downstream Processing
Gene Therapy Final Fill
Key considerations for viral vector production
Cell Therapy
Cell Therapy Upstream Processing
Cell Therapy Downstream Processing
Cell Therapy Final Fill
Promise and Potential: Cell Therapies for Pediatric Patients
Vaccines
Vaccines Upstream Processing
Vaccines Downstream Processing
Vaccines Formulation and Final Fill
Bioprocessing Solutions and Products for Vaccines
mRNA
Masterflex Peristaltic Pump and Fluid Handling Solutions
Upstream Bioprocess
Downstream Bioprocess
Formulation & Aseptic Fill-Finish
Life Sciences
Cell Biology
Cell Culture & Growth
Cellular Analysis
Genomics
qPCR
Gene Editing
Next Generation Sequencing
Proteomics
Protein Expression Technology
Protein Purification Methods
Protein Electrophoresis & Western Blotting
Microbiology
Featured Brands
Literature
Application Notes Library
Product Selector Guides
IgGy Antibody Selector
Chromatography
Chromatography for Pharma
Liquid Chromatography - Analytical
Column Selection
Avantor® HPLC and UHPLC columns and technical support
Chromatography for Biopharma
Large Molecule LC (Bio LC)
Chromatography for Food & Beverages
Chromatography for Environmental
Purification
Gas Chromatography
LC-MS
Liquid Chromatography - Sample Preparation
SPE Guide
Chromatography Application Library
Chromatography Educational Material
Chromatography Featured Brands
Quality Control & Testing
Atomic Spectroscopy
Electrochemistry
Molecular Spectroscopy
Physical and Chemical Properties
Environmental Testing
Titration
Chromatography
Mass Spectrometry
Quality Control and Testing Featured Brands
Quality Control and Testing Promotions
Quality Control and Testing Literature
Advanced Technologies & Applied Materials
Aviation and Defense
Space
Semiconductor and MEMS
Optoelectronics and Electronics
Food and Beverage
Controlled Environments
New Solutions
Solutions by Industry
Pharmaceuticals, Biotechnology and Nutraceuticals
Semiconductor and Microelectronics
Medical Device and Healthcare
Aerospace and Automotive
Controlled Environment & Cleanroom Sustainable Solutions
Sustainable Solutions
Sustainable Hygiene and Disposal Solutions
Greener Solutions for Controlled Environments
Recycling Solutions
Controlled Environment Featured Brands
Vendor Services
Controlled Environment Literature
Healthcare
Cytology
Blood Collection
Hematology
Histology
In Vitro Fertilization
Specimen Collection and Transport
Drug delivery
Medical implants
General healthcare
Healthcare Literature
Healthcare Featured Brands
Safety
Laboratory Safety
Manufacturing Personal Protection and Workwear
Facilities Maintenance, Protection and Hygiene
Safety Literature
Safety Featured Brands
Featured Solutions
Our Brands
J.T.Baker®
NuSil
Masterflex® Peristaltic Pump and Fluid Handling Solutions
VWR
Macron Fine Chemicals™
Featured Tools
Science Delivered
Supporting Research on COVID-19
J.T.Baker Robotic Tips
Lab Start-up Program
Our Collection of Brands
Research and Development
Equipment and Instrumentation
Ovens and Incubators
Centrifugation
Titration
Cold Storage
Weighing
Organic Synthesis
Spectrophotometers
Liquid Handling
Microscopy
R&D Literature
R&D Featured Brands
Production
Production Chemicals
Mining Supplies and Equipment
Sample Preparation
Fire Assay
Wet Chemistry
Metallurgical Testing
Refining
Environmental Testing
Chemicals
Safety
Weighing
Mining Literature
Science Education
Environmental Testing
Petrochemical
Standards and Methods
Food and Beverage
Autoclaves and Accessories
Cold Storage
Food Analysis
Rapid Microbiology Testing Kits
Sample Collection
Sample Preparation
Temperature & Humidify Measurement and Datalogging
Food and Beverage Literature
Food and Beverage Technical Articles
Academic Research
Lab Design & Furniture
VWR Rediship
Design and Plan Your Lab
Get Inspired
Lab Furniture
Lab Design and Furniture Featured Brands
Furniture Resources
BIMOS ergonomic chairs
Advanced Battery Science & Technology
Facility Management and Procurement
Metals, Mining and Refining
Battery Materials Production
Battery Cell, Module and Pack Assemblies
Battery Recycling
Battery R&D, QA/QC Testing
Chemicals and Raw Materials for Production
Advanced Battery Equipment and Instruments
Advanced Battery Personal Protective Equipment and Safety
Advanced Battery Lab Consumables
Advanced Battery Services
Support
Overview
Resources
Ask Avantor
Resources
Certificate of Analysis country codes
Grade Definitions
Silicone Definitions
Journey of Life Sciences
Literature
Promotions
Knowledge Center
Videos
Clearance Sale
Thought Leadership
Biopharma Thought Leadership
Healthcare Thought Leadeship
Advanced Technologies & Applied Materials Thought Leadership
Services Thought Leadership
Innovation & Quality Thought Leadership
Events
Webinars
Events List
ISO Certifications
Certificate/SDS Search
Local ISO Certifications
Working with Avantor
Managing Your Account
Shipping and Distribution
How To Ship
How To Sell
Help Center
Terms and Conditions
Global Contact
Local Contact
Literature
Promotions
New Lab Start-Up
About
Overview
Mission, Vision, Values
Leadership
Board of Directors
Scientific Advisory Board
History
Global Footprint
Custom Manufacturing
Innovation at Avantor
Behind the Innovation
Innovation in Biopharma
Bridgewater Innovation Center
Korea Innovation Center
Shanghai Innovation Center
Quality & Compliance
Management of Change
Local ISO Certifications
Sustainability
Sustainability Reports
People & Culture
Attracting & Developing Our Associates
Enhancing Our Culture of Belonging
Caring for Our Associates
Supplier Empowerment
Innovation & Environment
Operational Sustainability
Environmentally Preferred Products
Sustainable Product Innovation
CO2-Prestatieladder
Community Impact
Avantor Foundation
Associate Giving and Volunteerism
Governance & Integrity
Compliance & Certification
Responsible Supplier Program
Avantor Business System
Policies & statements
Our DEI Commitment
News
Careers
Investors
Global Footprint
Export
Orders Link
ContactUsLinkComponent
Not Found
Not Found | Avantor
Oops!
We couldn't find the page you are looking for.
Here are some useful links instead:
Home
FAQ