Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
POTASSIUM NITRATE 1 * 100 g
POTASSIUM NITRATE 1 * 100 g
Print
POTASSIUM NITRATE 1 * 100 g
Catalog # SIAL221295-100G
Supplier:
Merck
POTASSIUM NITRATE 1 * 100 g
Catalog # SIAL221295-100G
Supplier:
Merck
CAS Number:
Expand image
Return to Top
POTASSIUM NITRATE 1 * 100 g
Add to cart
CAS Number:
Price
39,00 €
Log in
for availability
Unit of Measure
1 * 100 g
-
+
Add to cart
Return Policy
Product Details & Documents