Manual Valves, CR4
CR4 valve, 2-way manual multi-turn, port 1 (inlet), port 2 (outlet) 1/4" Flaretek,
Catalog # ENTECR4-2M-4F
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
Smaller than most comparable valves, ¹/₄" CR4 series valves are ideal for handling wet etch and clean process chemicals. These durable valves can handle temperatures up to 160 °C at 276 kPa.
- Compact, modular design
- High-temperature capable, all-PFA construction
- High-purity corrosive chemical handling
- All semiconductor WEC process chemicals
- Chemical line sizes 3/8" or smaller
With no exposed metal hardware, the multi-turn valve is completely sealed and protected from harsh chemical environments. This all-PFA valve is available for high-purity chemical handling in line sizes 3/8" or smaller. A variety of connections including PrimeLock® , Flaretek® , FNPT, PureBond® , 'SpaceSaver', and Super 300 Type Pillar® allow flexibility in your fluid design. And, the compact, modular design enables manifold customisation.
Specifications
- Description:CR4 valve, 2-way manual multi-turn, port 1 (inlet), port 2 (outlet) 1/4" Flaretek,
- Seq:01