Screw Caps for Bottles
Original closure with cone, wide neck, GL 54, White, PP
Catalog # RIXI8-1110-54-W
Supplier: BERLIN PACKAGING RIXIUS GMBH
Jump to:
Product Details & Documents
Specifications
- Thread:GL 54
- Colour:White
- Description:Original closure with cone, wide neck
- Material:PP
- Seq:17