ContactUsLinkComponent
Specifications:- Capillary Column Phase: HP-5. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: 7 inch. In ner Diameter (ID): 0.32 mm. Length: 30 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 items
Catalog # HEWL19091J-413-KEY
Supplier:  Agilent
undefined
Specifications:- Capillary Column Phase: HP-5. Capillary Tubing: Fused Silica. Film Thickness: 0.25 um. GC Column Format: 7 inch. In ner Diameter (ID): 0.32 mm. Length: 30 m. Polarity: Low Polarity. With Smart Key: Yes. 1 * 1 items
Catalog # HEWL19091J-413-KEY
Supplier:  Agilent