96-well Deep-well plate, PP, V-bottom, well shape square, suitable for KingFisher™, 2,2 ml, CERTIFIED LIFE SCIENCE QUALITY 1 * 50 items
Catalog # BRND701348
96-well Deep-well plate, PP, V-bottom, well shape square, suitable for KingFisher™, 2,2 ml, CERTIFIED LIFE SCIENCE QUALITY 1 * 50 items
Catalog # BRND701348