Orders LinkContactUsLinkComponent
b.safe Desktop Distributors
b.safe Desktop Distributors
Desktop distributor, magnet attachment, incl. blind fittings (8× milky white, 4× white), fittings (8× green Ø 1,6 mm, 4× violet Ø 2,2 mm, 4× yellow Ø 3,2 mm), 8 capillary connectors, 4 hose connectors (Ø 6 - 8 mm, 90°), 1 straight hose connector (Ø 6 - 8 mm)
Catalog # BOHLR1122-04
Supplier:   Bohlender
b.safe Desktop Distributors
Catalog # BOHLR1122-04
Supplier:  Bohlender
b.safe Desktop Distributors
b.safe Desktop Distributors
Desktop distributor, magnet attachment, incl. blind fittings (8× milky white, 4× white), fittings (8× green Ø 1,6 mm, 4× violet Ø 2,2 mm, 4× yellow Ø 3,2 mm), 8 capillary connectors, 4 hose connectors (Ø 6 - 8 mm, 90°), 1 straight hose connector (Ø 6 - 8 mm)
Price
234,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Central waste collection from several HPLC systems. Capillaries and hoses are connected without kinks using the b.safe Fittings and b.safe Hose connectors included in the scope of delivery. The liquid solvent waste flows off directly and residue-free via the vertical hole inside the Desktop Distributor.
  • Central hose connection
  • Simple attachment to the work table
  • Residue-free drainage of solvent waste
Base plate made of PP, for collecting solvent waste from up to eight capillaries and four hoses and forwarding it via a central hose connection. With PFA-Fittings for capillaries with Ext.Ø 1, ⁶/₂, ²/₃, 2 mm (thread UNF ¹/₄"), tubing connectors for tubing Int.Ø 6 to 8 mm (thread NPT ¹/₈") as well as Blind Fittings for closing unused connections. Easy attachment to the worktable either by magnet or silicone-free adhesive pad. Design and scope of delivery according to table.
Specifications
  • Connection Type:
    8 capillary connectors, 4 hose connectors (Ø 6 - 8 mm, 90°), 1 straight hose connector (Ø 6 - 8 mm)
  • Description:
    Desktop distributor, magnet attachment, incl. blind fittings (8× milky white, 4× white), fittings (8× green Ø 1,6 mm, 4× violet Ø 2,2 mm, 4× yellow Ø 3,2 mm)
  • Seq:
    02