Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GC COLUMN HI-5MS 0.32MM 0.50UM 25M 1 * 1 items
Catalog # HIGCHI17-32-050-25
Supplier:
Avantor
Expand image
GC COLUMN HI-5MS 0.32MM 0.50UM 25M 1 * 1 items
Catalog # HIGCHI17-32-050-25
Supplier:
Avantor