Orders LinkContactUsLinkComponent
Nylon probe tools, Menda
Nylon probe, with fork blade point, 178 mm, Black
Catalog # 115-3064
Supplier:   DESCO
Nylon probe tools, Menda
Catalog # 115-3064
Supplier:  DESCO
Nylon probe tools, Menda
Nylon probe, with fork blade point, 178 mm, Black
Price
3,40 €
Log in for availability
1 * 1 items
Product Details & Documents
Ideal for opening plastic cases such as MP3 players, cell phones, laptops and more. Also a great tool for popping out batteries.
  • Nylon finish will not mark delicate surfaces
  • May be suitable for use in ESD protected areas
  • Temperature resistant
Specifications
  • Colour:
    Black
  • Description:
    Nylon probe, with fork blade point, 178 mm
  • Seq:
    01