Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Terpenes MegaMix Standard #1 (43 components), 2500 µg/mL in Isopropanol 1 * 1 mL
Catalog # REST34142
Supplier:
Restek
Expand image
Terpenes MegaMix Standard #1 (43 components), 2500 µg/mL in Isopropanol 1 * 1 mL
Catalog # REST34142
Supplier:
Restek