Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MDL 29951 1 * 5 mg
Catalog # DISX92-1264
Supplier:
DISCOVERX
Expand image
MDL 29951 1 * 5 mg
Catalog # DISX92-1264
Supplier:
DISCOVERX