Order Entry
Export
Orders LinkContactUsLinkComponent
Polysorbate 20, GPR RECTAPUR®

Polysorbate 20, GPR RECTAPUR®

Glass bottle, 100 mL
Catalog # 66368.180
Supplier:   VWR Chemicals
Polysorbate 20, GPR RECTAPUR®
Catalog # 66368.180
Supplier:  VWR Chemicals
CAS Number:  9005-64-5
Polysorbate 20, GPR RECTAPUR®
Polysorbate 20, GPR RECTAPUR®

Glass bottle, 100 mL

CAS Number:  9005-64-5
Price
28,20 €
Log in for availability
1 * 100 mL
Product Details & Documents

Tween® is a registered trade mark of Atlas Chemical Industries Inc.

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Density:1.1 g/cm³ (20 °C)
  • Flash Pt:149 °C (closed cup)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Appearance:
    Yellow to brown-yellow
  • IR Spectrum:
    Passes test
  • Density (20/4):
    1.095 - 1.105
  • Hydroxyl value:
    96 - 113
  • Saponification value:
    40 - 51

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications