Order Entry
Export
Orders LinkContactUsLinkComponent
PCRmax® Alpha Cyclers

PCRmax® Alpha Cyclers

Thermal cycler, single 384-well and single 96-well capacity, 100/240 VAC, 50/60 Hz
Catalog # TECN93945-20
Supplier:   Cole-Parmer
PCRmax® Alpha Cyclers
Catalog # TECN93945-20
Supplier:  Cole-Parmer
PCRmax® Alpha Cyclers
PCRmax® Alpha Cyclers

Thermal cycler, single 384-well and single 96-well capacity, 100/240 VAC, 50/60 Hz

Price
10 890,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Easily create, modify and access programs, saving time.

  • Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
  • Independently control each block for simplified temperature and space optimization
  • Software allows quick access to commonly used protocols

These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.

A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimise new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.

Specifications
  • Description:
    Thermal cycler, single 384-well and single 96-well capacity
  • Seq:
    06
  • Electrical supply:
    100/240 VAC, 50/60 Hz
  • Interfaces:
    USB
  • Programs:
    Max. 1000
  • Cat. No.:
    TECN93945-20
  • Gradient:
    +
  • Gradient temperature range:
    1...29 °C
  • Lid temperature range:
    35...115 °C, or off
  • Heating ramp rate:
    3,4 °C/s
  • Sample capacity:
    One 384-well plate, one set of 96×0,2 ml tubes or one 96-well plate
  • W×D×H:
    470×330×535 mm
  • Block homogeneity:
    ±0,3 at 55 °C
  • Temperature range:
    10...100 °C
  • Temperature accuracy:
    ±0,25 at 50 °C