PCRmax® Alpha Cyclers
Thermal cycler, single 384-well and triple 96-well capacity, 100/240 VAC, 50/60 Hz
Catalog # TECN93945-10
Supplier: Cole-Parmer
Jump to:
Product Details & Documents
Easily create, modify and access programs, saving time.
- Flexible—your choice of one, two, or four block capacities in 96-well, 384-well, or mixed formats
- Independently control each block for simplified temperature and space optimization
- Software allows quick access to commonly used protocols
These compact thermal cyclers deliver reproducible results every run—with little to no training needed. Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification.
A Program Wizard generates a protocol specific to your sequence in seconds—a quick way to optimise new assays. Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface. Save your personal settings to your USB drive; sign in is as simple as inserting your USB drive, no password needed.
Documents
Specifications
- Description:Thermal cycler, single 384-well and triple 96-well capacity
- Seq:07
- Electrical supply:100/240 VAC, 50/60 Hz
- Interfaces:USB
- Programs:Max. 1000
- Cat. No.:TECN93945-10
- Gradient:+
- Gradient temperature range:1...29 °C
- Lid temperature range:35...115 °C, or off
- Heating ramp rate:3,4 °C/s
- Sample capacity:One 384-well plate, three sets of 96×0,2 ml tubes or three 96-well plates
- W×D×H:700×330×535 mm
- Block homogeneity:±0,3 at 55 °C
- Temperature range:10...100 °C
- Temperature accuracy:±0,25 at 50 °C



