Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Plan Apochromat objective 100X/1.30/S-Oil (WD=0.25mm) 1 * 1 items
Catalog # MOTI1101001705781
Supplier:
MOTIC
Expand image
Plan Apochromat objective 100X/1.30/S-Oil (WD=0.25mm) 1 * 1 items
Catalog # MOTI1101001705781
Supplier:
MOTIC