Orders LinkContactUsLinkComponent
GloCell™ Fixable Viability Dye Violet 540
GloCell™ Fixable Viability Dye Violet 540
GloCell™ Fixable viability dye violet 540, 100 Tests
Catalog # STMC75011.1
GloCell™ Fixable Viability Dye Violet 540
Catalog # STMC75011.1
CAS Number:  
GloCell™ Fixable Viability Dye Violet 540
GloCell™ Fixable Viability Dye Violet 540
GloCell™ Fixable viability dye violet 540, 100 Tests
CAS Number:  
Price
184,00 €
Log in for availability
1 * 100 Tests
Product Details & Documents
Amine-labelling fluorescent dye for live/dead staining of mammalian cells - 100 tests.
GloCell™ Fixable viability dye violet 540 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be utilised prior to fixation, permeabilisation, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay.
GloCell™ Fixable viability dye violet 540 is excited by the violet laser at 405 nm and it has a fluorescence emission maximum of 540 nm that can be detected using the 525/50 band pass filter used to detect AmCyan.
CAS No: 67-68-5
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    100 Tests
  • Description:
    GloCell™ Fixable viability dye violet 540
Specifications