Order Entry
Export
Orders LinkContactUsLinkComponent
 

Human Recombinant FGFacidic, ACF

100 µG
Catalog # STMC78188.1
Human Recombinant FGFacidic, ACF
Catalog # STMC78188.1
CAS Number:  
Human Recombinant FGFacidic, ACF
Human Recombinant FGFacidic, ACF

100 µG

CAS Number:  
Price
462,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Fibroblast growth factor acidic (FGF-acidic), also known as FGF-1, is a potent activator of DNA synthesis, cell proliferation, and chemotaxis and is known to play numerous roles in development, regeneration, and angiogenesis (Galzie et al.; Jaye et al.; Presta et al.). FGF-acidic is produced by multiple cell types and is capable of activating all cells of mesodermal origin and many cells of neuroectodermal, ectodermal, and endodermal origin. It is found in large quantities in the brain, but is also expressed in hepatocytes, vascular smooth muscle cells, neurons of the central nervous system, skeletal muscle cells, fibroblasts, keratinocytes, endothelial cells, intestinal columnar epithelial cells, and pituitary basophils and acidophils. FGF-acidic is secreted as a disulfide-linked homodimer and is stored in complex with heparan sulfate, a requirement for its interaction with FGF receptors (Guerrini et al.; Mohammadi et al.). Internalized FGF-acidic signals via protein kinase C and promotes cell survival by inhibiting p53 and proapoptotic signaling (Bouleau et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    STMC78188.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FGFacidic, ACF
  • Purity:
    ≥95%
  • Species:
    Human

Specifications