Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant PDGF-BB, ACF

Human Recombinant PDGF-BB, ACF

10 µG
Catalog # STMC78154
Human Recombinant PDGF-BB, ACF
Catalog # STMC78154
CAS Number:  
Human Recombinant PDGF-BB, ACF
Human Recombinant PDGF-BB, ACF

10 µG

CAS Number:  
Price
310,00 €
Log in for availability
1 * 10 µG
Product Details & Documents

Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge-stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (Fretto et al.; Westermark and Heldin). PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta. PDGF-induced migration has been shown to involve MEK/ERK, EGFR, Src, and PI3K/Akt signaling pathways (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin- like fibroblasts, glial cells, and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). PDGF-BB is secreted by osteoblasts to induce mesenchymal stem cell migration and angiogenesis. It has also been shown that PDGF-BB is secreted by preosteoclasts during bone modeling and remodeling to induce angiogenesis and thus proper osteogenesis (Xie et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    STMC78154
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    PDGF-BB, ACF
  • Purity:
    ≥95%
  • Species:
    Human

Specifications