Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant IL-3, ACF

Human Recombinant IL-3, ACF

10 µG
Catalog # STMC78146
Human Recombinant IL-3, ACF
Catalog # STMC78146
CAS Number:  
Human Recombinant IL-3, ACF
Human Recombinant IL-3, ACF

10 µG

CAS Number:  
Price
329,00 €
Log in for availability
1 * 10 µG
Product Details & Documents

Interleukin 3 (IL-3) is a species-specific pleiotropic cytokine that promotes the survival and proliferation of pluripotent hematopoietic stem cells and lineage-committed progenitor cells and their differentiation into mature cells of most lineages, including basophils, neutrophils, eosinophils, macrophages, dendritic cells, erythrocytes, and megakaryocytes (Yang et al.; Dorssers et al.; Broughton et al.). IL-3 is produced by activated T cells and has a physiological role in inflammation and allergies by promoting the secretion of inflammatory mediators such as histamine, IL-4, and IL-6 by basophils and eosinophils (Broughton et al.). The IL-3 receptor consists of a unique alpha subunit (CD123) and a beta common subunit (βc or CD131) that is shared with the receptors for IL-5 and GM-CSF, and is the principal signal transduction subunit for these cytokines. IL-3 binding to the heterodimeric receptor activates JAK/STAT, MAPK, and PI3K signaling pathways (Woodcock et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    STMC78146
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-3, ACF
  • Purity:
    ≥95%
  • Species:
    Human

Specifications