Orders LinkContactUsLinkComponent
GloCell™ Fixable Viability Dye Red 780
GloCell™ Fixable Viability Dye Red 780
GloCell™ Fixable viability dye red 780, 500 tests, 500 Tests
Catalog # STMC75007
GloCell™ Fixable Viability Dye Red 780
Catalog # STMC75007
CAS Number:  
GloCell™ Fixable Viability Dye Red 780
GloCell™ Fixable Viability Dye Red 780
GloCell™ Fixable viability dye red 780, 500 tests, 500 Tests
CAS Number:  
Price
710,00 €
Log in for availability
1 * 500 Tests
Product Details & Documents
Amine-labelling fluorescent dye for live/dead staining of mammalian cells - 100 tests.
GloCell™ Fixable viability dye red 780 is a fluorescent cell viability dye for staining live/dead mammalian cells in applications such as flow cytometry. The dye irreversibly binds intracellular and cell surface amine groups and can be utilised prior to fixation, permeabilisation, or cryopreservation. Cells with compromised plasma membranes become permeable to the GloCell™ dye, resulting in a greater fluorescence compared to live cells in a cell viability assay.
GloCell™ Fixable viability dye red 780 is excited by the red laser at 633 nm and it has a fluorescence emission maximum of 780 nm that can be detected using the 780/60 band pass filter used to detect APC-Cy7.
CAS No: 67-68-5
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    500 Tests
  • Description:
    GloCell™ Fixable viability dye red 780
  • Pk Info:
    500 tests
Specifications