Order Entry
Export
Orders LinkContactUsLinkComponent
 

Cell Lysis Buffer, Abcam

Cell Lysis Buffer
Catalog # ABCAAB288311-400
Supplier:   Abcam
Cell Lysis Buffer, Abcam
Catalog # ABCAAB288311-400
Supplier:  Abcam
CAS Number:  
Cell Lysis Buffer, Abcam

Cell Lysis Buffer

CAS Number:  
Price
246,00 €
Log in for availability
1 * 400 mL
Product Details & Documents

This product is manufactured by BioVision, an Abcam company and was previously called 1067 Cell Lysis Buffer.

  • Constituents: 1% Alcohol Ethoxylate, 0.88% Sodium chloride, 0.32% Tris HCl

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Description:
    Cell Lysis Buffer
  • Cat. No.:
    ABCAAB288311-400
  • Storage temperature:
    Store at +4°C.

Specifications