Order Entry
Export
Orders LinkContactUsLinkComponent
Monochlorobimane (MCB) ≥98%

Monochlorobimane (MCB) ≥98%

Catalog # ABCAAB287129-5
Supplier:   Abcam
Monochlorobimane (MCB) ≥98%
Catalog # ABCAAB287129-5
Supplier:  Abcam
CAS Number:  76421-73-3
Monochlorobimane (MCB) ≥98%
CAS Number:  76421-73-3
Price
134,00 €
Log in for availability
1 * 5 mg
Product Details & Documents

Monochlorobimane is a cell-permeant probe for quantifying glutathione levels in cells. It is essentially nonfluorescent until it reacts with a thiol group. It readily reacts with several low molecular weight thiols, including cysteine, glutathione, N-acetylcysteine, mercaptopurine, peptides and plasma thiols. The glutathione conjugate of monochlorobimane has an excitation/emission maxima ~394/490 nM.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Chlorobimane, mBCI, 1H,7H-Pyrazolo(1,2-α)pyrazole-1,7-dione, 3-(chloromethyl)-2,5,6-trimethyl-
  • CAS Number:76421-73-3
  • Molecular Weight:226,66 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% 7-(chloromethyl)-1,2,6-trimethylpyrazolo[1,2-a]pyrazole-3,5-dione