Density Gradient Medium (Iodixanol)
Density Gradient Medium (Iodixanol)
Catalog # ABCAAB286850-250
Supplier: Abcam
Jump to:
Product Details & Documents
This product is manufactured by BioVision, an Abcam company and was previously called M1248 OptiPrep™ Density Gradient Medium.
- Constituents: 60% Iodixanol
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
- Description:Density Gradient Medium (Iodixanol)
- Cat. No.:ABCAAB286850-250
- Storage conditions:Store at Ambient.



