Order Entry
Export
Orders LinkContactUsLinkComponent
Ac-WEHD-AFC ≥97%

Ac-WEHD-AFC ≥97%

Catalog # ABCAAB285375-25
Supplier:   Abcam
Ac-WEHD-AFC ≥97%
Catalog # ABCAAB285375-25
Supplier:  Abcam
CAS Number:  
Ac-WEHD-AFC ≥97%
CAS Number:  
Price
769,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Ac-WEHD-AFC is a useful fluorogenic substrate for group I caspases (caspase-1, -4, and -5). Caspase activity can be quantified by fluorescent detection of free AFC that has an excitation maximum at 400 nm and emission maximum at 505 nm.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Caspase-1 substrate (fluorogenic), Caspase-5 substrate (fluorogenic)
  • Molecular Weight:838,7 g/mol