Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
EUGON BROTH - Dehydrated media - 500g 1 * 500 g
Catalog # BLIF4016432
Supplier:
BIOLIFE ITALIANA
Expand image
EUGON BROTH - Dehydrated media - 500g 1 * 500 g
Catalog # BLIF4016432
Supplier:
BIOLIFE ITALIANA