Orders LinkContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
P8x10, 3×4 measurements, 1 - 10 µl, ±0,08 - ±0,20 µl, ≤ 0,05 - ≤ 0,10 µl, Red, D10 / DL10 / DF10 / DFL10
Catalog # GILSF144068CG
Supplier:   GILSON
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Catalog # GILSF144068CG
Supplier:  GILSON
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
P8x10, 3×4 measurements, 1 - 10 µl, ±0,08 - ±0,20 µl, ≤ 0,05 - ≤ 0,10 µl, Red, D10 / DL10 / DF10 / DFL10
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling. The pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting.
  • Legendary performance
  • Easy pipetting
  • Built-to-last and convenient
  • Simple to maintain
Accurate and precise, PIPETMAN air-displacement pipettes are recognized as one of the world standard. They come in a large range of volumes, including single and multichannel models, allowing you to handle various sample types with confidence. PIPETMAN feels like PIPETMAN® Classic but with lower forces for enhanced pipetting habits with a standard pipette. Made of PVDF and stainless steel, PIPETMAN offers premium durability. Built-to-last, it is the economical solution for years of pipetting. Routine cleaning and maintenance are all that is required to keep PIPETMAN in top condition for years. The PIPETMAN design allows for easy access to user-serviceable parts, along with a fully autoclavable tip holder and ejector.
Ordering information: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories.
Specifications
  • Colour code:
    Red
  • Volume:
    1 - 10 µl
  • Description:
    P8x10, 3×4 measurements
  • Seq:
    09
  • Accuracy:
    ±0,08 - ±0,20 µl
  • Imprecision:
    ≤ 0,05 - ≤ 0,10 µl
  • Recommended tips:
    D10 / DL10 / DF10 / DFL10
Accessories
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)
    Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)
    612-1816
    White
    Log in for availability
    1 * 1 items - 126,00 €
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)
    Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)
    GILSF161405
    Translucent
    Log in for availability
    1 * 1 items - 111,00 €
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screws
    Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screws
    GILSF161406
    White
    Log in for availability
    1 * 1 items - 19,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Plastic ejector for P20
    Plastic ejector for P20
    GILSF107028
    Log in for availability
    1 * 1 items - 11,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Plastic ejector for P2/P10 with adapter
    Plastic ejector for P2/P10 with adapter
    GILSF107027
    Log in for availability
    1 * 1 items - 15,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Plastic ejector for P1000
    Plastic ejector for P1000
    GILSF107031
    Log in for availability
    1 * 1 items - 11,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Plastic ejector for P200
    Plastic ejector for P200
    GILSF107030
    Log in for availability
    1 * 1 items - 11,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Plastic ejector for P100
    Plastic ejector for P100
    GILSF107029
    Log in for availability
    1 * 1 items - 11,60 €