Micro soxhlet extraction apparatus, WHEATON®
Soxhlet extractor, 24-400 screw thread on bottom and 13-425 screw thread on side, — ml
Catalog # WHEA415501
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Used for continuous extraction of analytes from a solid into an organic solvent.
Specifications
- Description:Soxhlet extractor, 24-400 screw thread on bottom and 13-425 screw thread on side
- Seq:02
- Flask capacity:— ml