ContactUsLinkComponent
Transillumination Unit M LED - Lower part for stand M LED D340xW300xH58 mm - LED transillumination base with rotatable and slidable mirror for brightfield, darkfield and oblique transillumination - Plain mirror side for crisp, diffuse mirror side f - 1 * 1 ST
Catalog # ZEIS4355259100000
Supplier:  ZEISS
Transillumination Unit M LED - Lower part for stand M LED D340xW300xH58 mm - LED transillumination base with rotatable and slidable mirror for brightfield, darkfield and oblique transillumination - Plain mirror side for crisp, diffuse mirror side f - 1 * 1 ST
Catalog # ZEIS4355259100000
Supplier:  ZEISS