Micropump L24566 Pump Head Adapter Kit; NEMA 56C to A-Mount, High-Torque Mag
Catalog # MFLX07003-96
Supplier: Avantor Fluid Handling
About this item
Easily adapt your pump head to your drive with these adapter kits.
- Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.
- Description:Pump Head Adapter Kit; NEMA 56C to A-Mount, High-Torque Mag
- Environmentally Preferable:
- Pk:1