Pulsafeeder K4VTC1 Service Kit for Solenoid Metering Pump, 21- 44 GPD
Catalog # MFLX78118-82
Supplier: Avantor Fluid Handling
About this item
Optimize use of your Pulsafeeder metering pumps.
- Service kits are suitable for use with Pulsafeeder solenoid metering pumps
Replace wearable parts to extend the life of your pump.
- Description:Service Kit for Solenoid Metering Pump, 21- 44 GPD
- Environmentally Preferable:
- Warranty:3 Months
- Pk:1